1-Nonanesulfonic acid sodium salt 98+%
Supplier: Thermo Scientific Chemicals
L06407-06
L06407-14
CAAAAL06407-06EA
131.77
CAD
CAAAAL06407-06
CAAAAL06407-14
1-Nonanesulfonic acid sodium salt 98+%
1-Nonanesulphonic acid sodium salt
Purity: 98+%
CAS No.: 35192-74-6
Molecular Formula: C9H19NaO3S
Formula Weight: 230.30
Melting Point:
Formula:
C₉H₁₉NaO₃S Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00025010 CAS Number: 35192-74-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...