4-tert-Butylbenzophenone 98%
Supplier: Thermo Scientific Chemicals
L06386-03
L06386-06
CAAAAL06386-03EA
66.2
CAD
CAAAAL06386-03
CAAAAL06386-06
4-tert-Butylbenzophenone 98%
4-tert-Butylbenzophenone
Purity: 98%
CAS No.: 22679-54-5
Molecular Formula: C17H18O
Formula Weight: 238.32
Flash Point: 150°(302°F)
Melting Point: 38-42°
Boiling Point: 168-170°/2mm
Beilstein Reference: 2100075
MDL No.: MFCD00017255
Formula:
C₁₇H₁₈O Boiling Pt: 168…170 °C (2 mmHg) Melting Pt: 38…42 °C Flash Pt: 150 °C (302 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00017255 CAS Number: 22679-54-5 EINECS: 245-154-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...