Glycolaldehyde diethyl acetal 98% stabilized
Supplier: Thermo Scientific Chemicals
Warning
|
Stabilization:
Stabilised with sodium hydroxide 0.1%
L06282-04
L06282-09
L06282-18
CAAAAL06282-04EA
85.16
CAD
CAAAAL06282-04
CAAAAL06282-09
CAAAAL06282-18
Glycolaldehyde diethyl acetal 98% stabilized
Glycolaldehyde diethyl acetal
Stab. with ca 0.1% sodium carbonate, 98%
Purity: 98%
CAS No.: 621-63-6
Molecular Formula: HOCH2CH(OCH2CH3)2
Formula Weight: 134.18
Storage Temperature: 2°C to 8°C
Flash Point: 62°(143°F)
Refractive Index: 1.4160
Boiling Point: 166-167°
Beilstein Reference: 773727
MDL No.: MFCD00051486
Formula:
HOCH₂CH(OCH₂CH₃)₂ Boiling Pt: 166…167 °C Flash Pt: 62 °C(143 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00051486 CAS Number: 621-63-6 EINECS: 210-697-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...