Tris(trimethylsilyl)borate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L06277-06
L06277-14
CAAAAL06277-06EA
46.45
CAD
CAAAAL06277-06
CAAAAL06277-14
Tris(trimethylsilyl)borate 98%
Tris(trimethylsilyl) borate
Purity: 98%
CAS No.: 4325-85-3
Molecular Formula: [(CH3)3SiO]3B
Formula Weight: 278.38
Flash Point: 42°(107°F)
Refractive Index: 1.3861
Melting Point: -35°
Boiling Point: 183-185°
Beilstein Reference: 1772733
UN No.: 1993
MDL No.: MFCD00051588
Formula:
C₉H₂₇BO₃Si₃ Boiling Pt: 183…185 °C Melting Pt: –35 °C Density: 0.834 Flash Pt: 42 °C(107 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00051588 CAS Number: 4325-85-3 UN: 1993 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...