Piperonylacetone 98%

Supplier: Thermo Scientific Chemicals

L06258-06 L06258-14
CAAAAL06258-06EA 78.37 CAD
CAAAAL06258-06 CAAAAL06258-14
Piperonylacetone 98%
Piperonylacetone

Purity: 98% CAS No.: 55418-52-5 Molecular Formula: C11H12O3 Formula Weight: 192.22 Flash Point: 110°(230°F) Melting Point: 49-53° Boiling Point: 164-165°/12mm Beilstein Reference: 169843 MDL No.: MFCD00016910

Formula: C₁₁H₁₂O₃
Boiling Pt: 164…165 °C/12 mm
Melting Pt: 49…53 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00016910
CAS Number: 55418-52-5
EINECS: 259-630-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR