3-(2-Chlorophenyl)-5-methylisoxazole-4-carboxylic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L06240-09
L06240-18
L06240-30
CAAAAL06240-09EA
46.77
CAD
CAAAAL06240-09
CAAAAL06240-18
CAAAAL06240-30
3-(2-Chlorophenyl)-5-methylisoxazole-4-carboxylic acid 98%
3-(2-Chlorophenyl)-5-methylisoxazole-4-carboxylic acid
Purity: 98+%
CAS No.: 23598-72-3
Molecular Formula: C11H8ClNO3
Formula Weight: 237.64
Melting Point: 187-192°
Beilstein Reference: 791928
Formula:
C₁₁H₈ClNO₃ Melting Pt: 187…192 °C Storage Temperature: Ambient |
MDL Number:
MFCD00020813 CAS Number: 23598-72-3 EINECS: 245-770-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...