N-(2-Chloroethyl)benzamide 97%
Supplier: Thermo Scientific Chemicals
L06212-09
L06212-18
CAAAAL06212-09EA
55.77
CAD
CAAAAL06212-09
CAAAAL06212-18
N-(2-Chloroethyl)benzamide 97%
N-(2-Chloroethyl)benzamide
Purity: 97%
CAS No.: 26385-07-9
Molecular Formula: C9H10ClNO
Formula Weight: 183.64
Melting Point: 101-105°
Beilstein Reference: 1943939
MDL No.: MFCD00000964
Formula:
C₉H₁₀ClNO MW: 183.635 g/mol Melting Pt: 101…105 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000964 CAS Number: 26385-07-9 EINECS: 247-641-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...