Methyl-2-nitro-3,4,5-trimethoxybenzoate 98%

Supplier: Thermo Scientific Chemicals
Warning

L06188-09 L06188-18
CAAAAL06188-09EA 40.76 CAD
CAAAAL06188-09 CAAAAL06188-18
Methyl-2-nitro-3,4,5-trimethoxybenzoate 98%
Methyl-2-nitro-3,4,5-trimethoxybenzoate

Purity: 98% CAS No.: 5081-42-5 Molecular Formula: C11H13NO7 Formula Weight: 271.23 Melting Point: 66-70° Beilstein Reference: 2702653 MDL No.: MFCD00007097

Formula: C₁₁H₁₃NO₇
MW: 271.23 g/mol
Melting Pt: 66…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00007097
CAS Number: 5081-42-5
EINECS: 225-794-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR