4-Methoxybenzoic anhydride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L06180-06
L06180-14
CAAAAL06180-06EA
110.76
CAD
CAAAAL06180-06
CAAAAL06180-14
4-Methoxybenzoic anhydride 98%
4-Methoxybenzoic anhydride
Purity: 98%
CAS No.: 794-94-5
Molecular Formula: C16H14O5
Formula Weight: 286.29
Melting Point: 95-99°
Beilstein Reference: 545712
Formula:
C₁₆H₁₄O₅ Melting Pt: 95…99 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017175 CAS Number: 794-94-5 EINECS: 212-345-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...