trans-1-Methoxy-3-trimethylsilyloxy-1,3-butadiene 94%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L06100-03
L06100-06
CAAAAL06100-03EA
87.22
CAD
CAAAAL06100-03
CAAAAL06100-06
trans-1-Methoxy-3-trimethylsilyloxy-1,3-butadiene 94%
trans-1-Methoxy-3-trimethylsilyloxy-1,3-butadiene
Fieser: 6,370 7,233 9,303 10,260 11,332 12,318 16,163 17,182 18,236
Purity: 94%
CAS No.: 54125-02-9
Molecular Formula: C8H16O2Si
Formula Weight: 172.30
Storage Temperature: 2°C to 8°C
Flash Point: 45°(113°F)
Refractive Index: 1.4555
Boiling Point: 68-69°/14mm
Beilstein Reference: 1616761
UN No.: 1993
MDL No.: MFCD00008498
Formula:
C₈H₁₆O₂Si Boiling Pt: 68…69 °C/14 mm Density: 0.890 Flash Pt: 45 °C (113 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00008498 CAS Number: 54125-02-9 UN: 1993 ADR: 3,III Merck Index: 14,6001 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...