2-(4-Chlorophenyl)propionic acid 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L06020-06
L06020-14
CAAAAL06020-06EA
142.83
CAD
CAAAAL06020-06
CAAAAL06020-14
2-(4-Chlorophenyl)propionic acid 97%
2-(4-Chlorophenyl)propionic acid
Purity: 97%
CAS No.: 938-95-4
Molecular Formula: C9H9ClO2
Formula Weight: 184.62
Melting Point: 55-59°
Boiling Point: 121-126°/0.2mm
Beilstein Reference: 2328710
MDL No.: MFCD00044670
Formula:
C₉H₉ClO₂ Boiling Pt: 121…126 °C/0.2 mm Melting Pt: 55…59 °C Storage Temperature: Ambient |
MDL Number:
MFCD00044670 CAS Number: 938-95-4 EINECS: 213-351-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...