4'-Chloro-3'-methylacetophenone ≥75% remainder 3'-chloro-4'-methylacetophenone, tech.

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(4-Chloro-3-methylphenyl)ethanone

L06011-06 L06011-14
CAAAAL06011-06EA 99.86 CAD
CAAAAL06011-06 CAAAAL06011-14
4'-Chloro-3'-methylacetophenone ≥75% remainder 3'-chloro-4'-methylacetophenone, tech.
4'-Chloro-3'-methylacetophenone

Purity: 75% CAS No.: 37074-39-8 Molecular Formula: C9H9ClO Formula Weight: 168.62 Flash Point: 110°(230°F) Boiling Point: 104-105°/0.4mm MDL No.: MFCD00045172

Formula: C₉H₉ClO
MW: 168.62 g/mol
Boiling Pt: 104…105 °C/0.4 mm
Density: 1,15
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00045172
CAS Number: 37074-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR