Methyl-4,4-dimethyl-3-oxovalerate 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L06007-06
L06007-14
CAAAAL06007-06EA
81.21
CAD
CAAAAL06007-06
CAAAAL06007-14
Methyl-4,4-dimethyl-3-oxovalerate 95%
Methyl-4,4-dimethyl-3-oxovalerate
Purity: 95%
CAS No.: 55107-14-7
Molecular Formula: C8H14O3
Formula Weight: 158.20
Flash Point: 77°(170°F)
Refractive Index: 1.4320
Boiling Point: 67-70°/13mm
Beilstein Reference: 1764796
MDL No.: MFCD00008847
Formula:
C₈H₁₄O₃ Boiling Pt: 67…70 °C/13 mm Density: 0.990 Flash Pt: 77 °C(170 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008847 CAS Number: 55107-14-7 EINECS: 259-481-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...