6-Chloropiperonyl alcohol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L05972-06
L05972-14
CAAAAL05972-06EA
65.73
CAD
CAAAAL05972-06
CAAAAL05972-14
6-Chloropiperonyl alcohol 98%
6-Chloropiperonyl alcohol
Purity: 98%
CAS No.: 2591-25-5
Molecular Formula: C8H7ClO3
Formula Weight: 186.60
Melting Point: 68-72°
Beilstein Reference: 150801
MDL No.: MFCD00051727
Formula:
C₈H₇ClO₃ Melting Pt: 68…72 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051727 CAS Number: 2591-25-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...