L-Arginine methyl ester dihydrochloride 98%
Supplier: Thermo Scientific Chemicals
L05959-06
L05959-14
CAAAAL05959-06EA
57.51
CAD
CAAAAL05959-06
CAAAAL05959-14
L-Arginine methyl ester dihydrochloride 98%
L-Arginine methyl ester dihydrochloride
Purity: 98%
CAS No.: 26340-89-6
Molecular Formula: C7H16N4O2.2HCl
Formula Weight: 261.15
Melting Point: ca 190° dec.
Beilstein Reference: 4159929
Optical Rotation: +21° (c=2.5 in methanol)
MDL No.: MFCD00038948
Formula:
C₇H₁₆N₄O₂ Melting Pt: ca 190 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00038948 CAS Number: 26340-89-6 EINECS: 247-622-0 |
Specification Test Results
Optical rotation | +21° (c=2.5 in methanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...