1-Methylpyrrole-2-carboxylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Methyl-1H-pyrrole-2-carboxylic acid, 1-Methyl-2-pyrrolecarboxylic acid

L05906-03 L05906-06
CAAAAL05906-03EA 53.88 CAD
CAAAAL05906-03 CAAAAL05906-06
1-Methylpyrrole-2-carboxylic acid 98%
1-Methylpyrrole-2-carboxylic acid

Purity: 98% CAS No.: 6973-60-0 Molecular Formula: C6H7NO2 Formula Weight: 125.13 Melting Point: 135-138° Beilstein Reference: 115703

Formula: C₆H₇NO₂
MW: 125.13 g/mol
Melting Pt: 135…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00003088
CAS Number: 6973-60-0
EINECS: 230-217-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR