2-Butyn-1-al diethyl acetal 98%

Supplier: Thermo Scientific Chemicals
Warning

L05872-03 L05872-06
CAAAAL05872-03EA 44.24 CAD
CAAAAL05872-03 CAAAAL05872-06 CAAAAL05872-14
2-Butyn-1-al diethyl acetal 98%
2-Butyn-1-al diethyl acetal

Purity: 98% CAS No.: 2806-97-5 Molecular Formula: CH3C≡CCH(OCH2CH3)2 Formula Weight: 142.20 Flash Point: 56°(133°F) Refractive Index: 1.4260 Odor: Recognizable Boiling Point: 65-67°/12mm Beilstein Reference: 1702075 UN No.: 1993 MDL No.: MFCD00009274

Formula: C₈H₁₄O₂
Boiling Pt: 65…67 °C/12 mm
Density: 0.898
Flash Pt: 56 °C(133 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009274
CAS Number: 2806-97-5
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR