O-(4-Nitrobenzyl)hydroxylamine hydrochloride 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
O-(4-Nitrobenzyl)-hydroxylammonium chloride
L05861-03
L05861-06
CAAAAL05861-03EA
107.44
CAD
CAAAAL05861-03
CAAAAL05861-06
O-(4-Nitrobenzyl)hydroxylamine hydrochloride 98%
O-(4-Nitrobenzyl)hydroxylamine hydrochloride
Purity: 98%
CAS No.: 2086-26-2
Molecular Formula: C7H8N2O3.HCl
Formula Weight: 204.61
Odor: Nearly odorless
Melting Point: ca 215° dec.
Beilstein Reference: 3709565
MDL No.: MFCD00012954
Formula:
C₇H₉ClN₂O₃ Melting Pt: ca 215 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012954 CAS Number: 2086-26-2 EINECS: 218-228-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...