3-Indazolinone 97%
Supplier: Thermo Scientific Chemicals
L05860-06
L05860-14
L05860-22
CAAAAL05860-06EA
125.29
CAD
CAAAAL05860-06
CAAAAL05860-14
CAAAAL05860-22
3-Indazolinone 97%
3-Indazolinone
Purity: 97%
CAS No.: 7364-25-2
Molecular Formula: C7H6N2O
Formula Weight: 134.14
Melting Point: 248-252°
Beilstein Reference: 3733
MDL No.: MFCD00005685
Formula:
C₇H₆N₂O Melting Pt: 248…252 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005685 CAS Number: 7364-25-2 EINECS: 230-904-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...