Benzo[b]thiophene-3-acetic acid ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(Benzo[b]thiophen-3-yl)acetic acid
L05855-04
L05855-09
L05855-ME
CAAAAL05855-04EA
380.78
CAD
CAAAAL05855-04
CAAAAL05855-09
CAAAAL05855-ME
Benzo[b]thiophene-3-acetic acid ≥98%
Benzo[b]thiophene-3-acetic acid
Purity: 98+%
CAS No.: 1131-09-5
Molecular Formula: C10H8O2S
Formula Weight: 192.24
Melting Point: 108-112°
Beilstein Reference: 143357
MDL No.: MFCD00051637
Formula:
C₁₀H₈O₂S Melting Pt: 108…112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051637 CAS Number: 1131-09-5 EINECS: 214-461-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...