2-Amino-4-imino-2-thiazoline hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Warning

L05844-06 L05844-14
CAAAAL05844-06EA 28.28 CAD
CAAAAL05844-06 CAAAAL05844-14
2-Amino-4-imino-2-thiazoline hydrochloride 99%
2-Amino-4-imino-2-thiazoline hydrochloride

Purity: 99% CAS No.: 36518-76-0 Molecular Formula: C3H5N3S.HCl Formula Weight: 151.62 Melting Point: 300° Beilstein Reference: 5109728 MDL No.: MFCD00012721

Formula: C₃H₅N₃S·HCl
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00012721
CAS Number: 36518-76-0
EINECS: 253-083-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR