2,5-Dimethyl-1-phenylpyrrole-3-carboxaldehyde ≥98%
Supplier: Thermo Scientific Chemicals
L05836-06
L05836-14
CAAAAL05836-06EA
156.1
CAD
CAAAAL05836-06
CAAAAL05836-14
2,5-Dimethyl-1-phenylpyrrole-3-carboxaldehyde ≥98%
2,5-Dimethyl-1-phenylpyrrole-3-carboxaldehyde
Purity: 98+%
CAS No.: 83-18-1
Molecular Formula: C13H13NO
Formula Weight: 199.25
Storage Temperature: 2°C to 8°C
Melting Point: 87-90°
Beilstein Reference: 144790
MDL No.: MFCD00051494
Formula:
C₁₃H₁₃NO Melting Pt: 87…90 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00051494 CAS Number: 83-18-1 EINECS: 201-458-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...