1,2-Ethylenediphosphonic acid 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L05828-06
L05828-14
CAAAAL05828-06EA
195.92
CAD
CAAAAL05828-06
CAAAAL05828-14
1,2-Ethylenediphosphonic acid 97%
1,2-Ethylenediphosphonic acid
Purity: 97%
CAS No.: 6145-31-9
Molecular Formula: C2H8O6P2
Formula Weight: 190.03
Melting Point: 217-221°
Beilstein Reference: 1780250
UN No.: 3261
MDL No.: MFCD00013958
Formula:
C₂H₈O₆P₂ Melting Pt: 217…221 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013958 CAS Number: 6145-31-9 UN: 3261 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...