Ethyl vanillate 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Ethyl 4-hydroxy-3-methoxybenzoate

L05798-09 L05798-18
CAAAAL05798-09EA 93.06 CAD
CAAAAL05798-09 CAAAAL05798-18
Ethyl vanillate 97%
Ethyl vanillate

Purity: 97% CAS No.: 617-05-0 Molecular Formula: C10H12O4 Formula Weight: 196.20 Melting Point: 42-46° Boiling Point: 293° MDL No.: MFCD00017269

Formula: C₁₀H₁₂O₄
MW: 196.20 g/mol
Boiling Pt: 293 °C
Melting Pt: 42…46 °C
Storage Temperature: Ambient
MDL Number: MFCD00017269
CAS Number: 617-05-0
EINECS: 210-503-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR