4-(Trifluoromethyl)benzoic acid hydrazide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L05724-03
L05724-06
CAAAAL05724-03EA
40.29
CAD
CAAAAL05724-03
CAAAAL05724-06
4-(Trifluoromethyl)benzoic acid hydrazide 98%
4-(Trifluoromethyl)benzoic acid hydrazide
Purity: 98%
CAS No.: 339-59-3
Molecular Formula: C8H7F3N2O
Formula Weight: 204.15
Melting Point: 117-121°
Beilstein Reference: 1968848
Formula:
C₈H₇F₃N₂O Melting Pt: 117…121 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051703 CAS Number: 339-59-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...