5-Hydroxy-3,4-dimethoxybenzoic acid ≥97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L05708-03
L05708-MD
CAAAAL05708-03EA
486.64
CAD
CAAAAL05708-03
CAAAAL05708-MD
5-Hydroxy-3,4-dimethoxybenzoic acid ≥97%
5-Hydroxy-3,4-dimethoxybenzoic acid
Purity: 97+%
CAS No.: 1916-08-1
Molecular Formula: C9H10O5
Formula Weight: 198.17
Melting Point: 194-197°
Beilstein Reference: 2115310
MDL No.: MFCD00002508
Formula:
C₉H₁₀O₅ MW: 198.17 g/mol Melting Pt: 194…197 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002508 CAS Number: 1916-08-1 EINECS: 217-630-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...