4-Chloro-1-methylpiperidine hydrochloride 97%
Supplier: Thermo Scientific Chemicals
L05699-14
L05699-22
CAAAAL05699-14EA
41.87
CAD
CAAAAL05699-14
CAAAAL05699-22
4-Chloro-1-methylpiperidine hydrochloride 97%
4-Chloro-1-methylpiperidine hydrochloride
Purity: 97%
CAS No.: 5382-23-0
Molecular Formula: C6H12ClN.HCl
Formula Weight: 170.08
Melting Point: 159-166°
Beilstein Reference: 3677071
Formula:
C₆H₁₃Cl₂N MW: 170.08 g/mol Melting Pt: 159…166 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012834 CAS Number: 5382-23-0 EINECS: 226-375-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...