Methyl-4-(methylsulfonylmethyl)benzoate 98%
Supplier: Thermo Scientific Chemicals
L05657-04
L05657-09
CAAAAL05657-04EA
113.13
CAD
CAAAAL05657-04
CAAAAL05657-09
Methyl-4-(methylsulfonylmethyl)benzoate 98%
Methyl-4-(methylsulfonylmethyl)benzoate
Purity: 98%
CAS No.: 160446-22-0
Molecular Formula: C10H12O4S
Formula Weight: 228.27
Melting Point: 164-166°
Beilstein Reference: 7810971
MDL No.: MFCD00051693
Formula:
C₁₀H₁₂O₄S MW: 228.27 g/mol Melting Pt: 164…166 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051693 CAS Number: 160446-22-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...