2-(4-Ethoxyphenyl)ethanol 98%
Supplier: Thermo Scientific Chemicals
L05597-04
L05597-09
CAAAAL05597-04EA
94.01
CAD
CAAAAL05597-04
CAAAAL05597-09
2-(4-Ethoxyphenyl)ethanol 98%
2-(4-Ethoxyphenyl)ethanol
Purity: 98%
CAS No.: 22545-15-9
Molecular Formula: C10H14O2
Formula Weight: 166.22
Melting Point: 40-44°
Boiling Point: 138-141°/8mm
Beilstein Reference: 3239377
MDL No.: MFCD00016570
Formula:
C₁₀H₁₄O₂ Boiling Pt: 138…141 °C/8 mm Melting Pt: 40…44 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016570 CAS Number: 22545-15-9 EINECS: 245-072-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...