2-Decyn-1-ol 97%

Supplier: Thermo Scientific Chemicals

L05523-06 L05523-14
CAAAAL05523-06EA 95.12 CAD
CAAAAL05523-06 CAAAAL05523-14
2-Decyn-1-ol 97%
2-Decyn-1-ol

Purity: 97% CAS No.: 4117-14-0 Molecular Formula: CH3(CH2)6C≡CCH2OH Formula Weight: 154.25 Flash Point: 110°(230°F) Refractive Index: 1.4585 Boiling Point: 114-116°/6mm Beilstein Reference: 1852842 MDL No.: MFCD00041567

Formula: C₁₀H₁₈O
Boiling Pt: 114…116 °C/6 mm
Density: 0.855
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041567
CAS Number: 4117-14-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR