2-(4-Methoxybenzoyl)thiophene 97%

Supplier: Thermo Scientific Chemicals

L05513-09 L05513-18
CAAAAL05513-09EA 122.77 CAD
CAAAAL05513-09 CAAAAL05513-18
2-(4-Methoxybenzoyl)thiophene 97%
2-(4-Methoxybenzoyl)thiophene

Purity: 97% CAS No.: 4160-63-8 Molecular Formula: C12H10O2S Formula Weight: 218.27 Melting Point: 71-75° Beilstein Reference: 148560 MDL No.: MFCD00005440

Formula: C₁₂H₁₀O₂S
Melting Pt: 71…75 °C
Storage Temperature: Ambient
MDL Number: MFCD00005440
CAS Number: 4160-63-8
EINECS: 223-997-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR