4-Chloro-1,8-naphthalic anhydride 94%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L05508-14
L05508-22
L05508-36
CAAAAL05508-14EA
66.52
CAD
CAAAAL05508-14
CAAAAL05508-22
CAAAAL05508-36
4-Chloro-1,8-naphthalic anhydride 94%
4-Chloro-1,8-naphthalic anhydride
Purity: 94%
CAS No.: 4053-08-1
Molecular Formula: C12H5ClO3
Formula Weight: 232.63
Melting Point: 208-214°
Beilstein Reference: 178072
Formula:
C₁₂H₅ClO₃ Melting Pt: 208…214 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006928 CAS Number: 4053-08-1 EINECS: 223-760-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...