3,4-Dimethoxyphenylacetone 97%
Supplier: Thermo Scientific Chemicals
L05477-09
L05477-18
CAAAAL05477-09EA
42.5
CAD
CAAAAL05477-09
CAAAAL05477-18
3,4-Dimethoxyphenylacetone 97%
3,4-Dimethoxyphenylacetone
Purity: 97%
CAS No.: 776-99-8
Molecular Formula: C11H14O3
Formula Weight: 194.23
Flash Point:
Formula:
C₁₁H₁₄O₃ Boiling Pt: 154…158 °C/12 mm Density: 1,115 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008772 CAS Number: 776-99-8 EINECS: 212-285-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...