1-Methyl-3-n-propyl-2-pyrazolin-5-one 97%
Supplier: Thermo Scientific Chemicals
L05425-03
L05425-06
CAAAAL05425-03EA
45.19
CAD
CAAAAL05425-03
CAAAAL05425-06
1-Methyl-3-n-propyl-2-pyrazolin-5-one 97%
1-Methyl-3-n-propyl-2-pyrazolin-5-one
Purity: 97%
CAS No.: 31272-04-5
Molecular Formula: C7H12N2O
Formula Weight: 140.19
Melting Point: 114-118°
Beilstein Reference: 637013
MDL No.: MFCD00051849
Formula:
C₇H₁₂N₂O Melting Pt: 114…118 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051849 CAS Number: 31272-04-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...