Ethyl-4-hydroxymandelate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L05413-03
L05413-06
CAAAAL05413-03EA
49.77
CAD
CAAAAL05413-03
CAAAAL05413-06
Ethyl-4-hydroxymandelate 98%
Ethyl-4-hydroxymandelate
Purity: 98%
CAS No.: 68758-68-9
Molecular Formula: C10H12O4
Formula Weight: 196.20
Melting Point: 128-132°
Beilstein Reference: 3132065
MDL No.: MFCD00020180
Formula:
C₁₀H₁₂O₄ Melting Pt: 128…132 °C Storage Temperature: Ambient |
MDL Number:
MFCD00020180 CAS Number: 68758-68-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...