(3-Cyanopropyl)triethoxysilane 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(Triethoxysilyl)butanenitrile

L05367-14 L05367-22
CAAAAL05367-14EA 75.37 CAD
CAAAAL05367-14 CAAAAL05367-22
(3-Cyanopropyl)triethoxysilane 97%
(3-Cyanopropyl)triethoxysilane

Purity: 97% CAS No.: 1067-47-6 Molecular Formula: C9H19NO3 Formula Weight: 231.37 Flash Point: 74°(165°F) Refractive Index: 1.4170 Boiling Point: 238-241° Beilstein Reference: 1942237 MDL No.: MFCD00042636

Formula: C₉H₁₉NO₃
MW: 231.37 g/mol
Boiling Pt: 238…241 °C
Density: 0.961
Flash Pt: 74 °C(165 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042636
CAS Number: 1067-47-6
EINECS: 213-931-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR