1-Phenyl-1-(trimethylsilyloxy)ethylene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L05308-03
L05308-06
CAAAAL05308-03EA
46.77
CAD
CAAAAL05308-03
CAAAAL05308-06
1-Phenyl-1-(trimethylsilyloxy)ethylene 97%
1-Phenyl-1-(trimethylsilyloxy)ethylene
Purity: 97%
CAS No.: 13735-81-4
Molecular Formula: C11H16OSi
Formula Weight: 192.34
Flash Point: 78°(172°F)
Refractive Index: 1.5020
Boiling Point: 53-54°/1mm
Beilstein Reference: 1306914
MDL No.: MFCD00008582
Formula:
C₁₁H₁₆OSi Boiling Pt: 53…54 °C/1 mm Density: 0,938 Flash Pt: 78 °C(172 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00008582 CAS Number: 13735-81-4 EINECS: 237-308-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...