N,N-Bis(2-chloroethyl)-p-toluenesulfonamide 90%, tech.
Supplier: Thermo Scientific Chemicals
|
Warning
|
L05296-06
L05296-14
CAAAAL05296-06EA
61.62
CAD
CAAAAL05296-06
CAAAAL05296-14
N,N-Bis(2-chloroethyl)-p-toluenesulfonamide 90%, tech.
N,N-Bis(2-chloroethyl)-p-toluenesulfonamide
Purity: 90%
CAS No.: 42137-88-2
Molecular Formula: C11H15Cl2NO2S
Formula Weight: 296.22
Melting Point: 45-47°
Beilstein Reference: 2134258
MDL No.: MFCD00018944
Formula:
C₁₁H₁₅Cl₂NO₂S Melting Pt: 45…47 °C Storage Temperature: Ambient |
MDL Number:
MFCD00018944 CAS Number: 42137-88-2 EINECS: 255-677-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...