4-Chloro-α,α-dimethylphenylacetic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L05277-03
L05277-06
CAAAAL05277-03EA
28.91
CAD
CAAAAL05277-03
CAAAAL05277-06
4-Chloro-α,α-dimethylphenylacetic acid 98%
4-Chloro-alpha,alpha-dimethylphenylacetic acid
Purity: 98%
CAS No.: 6258-30-6
Molecular Formula: C10H11ClO2
Formula Weight: 198.65
Melting Point: 124-129°
Beilstein Reference: 2094056
MDL No.: MFCD00044814
Formula:
C₁₀H₁₁ClO₂ Melting Pt: 124…129 °C Storage Temperature: Ambient |
MDL Number:
MFCD00044814 CAS Number: 6258-30-6 EINECS: 228-392-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...