Diethyl (4-methoxybenzyl)phosphonate 98+%

Supplier: Thermo Scientific Chemicals

L05226-09 L05226-18
CAAAAL05226-09EA 115.34 CAD
CAAAAL05226-09 CAAAAL05226-18
Diethyl (4-methoxybenzyl)phosphonate 98+%
Diethyl (4-methoxybenzyl)phosphonate

Purity: 98+% CAS No.: 1145-93-3 Molecular Formula: C12H19O4P Formula Weight: 258.25 Flash Point: 110°(230°F) Refractive Index: 1.5045 Boiling Point: 136-138°/0.7mm Beilstein Reference: 2125148 MDL No.: MFCD00015662

Formula: C₁₂H₁₉O₄P
Boiling Pt: 136…138 °C/0.7 mm
Density: 1,500
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015662
CAS Number: 1145-93-3
EINECS: 214-548-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR