11-Heneicosanol 97%
Supplier: Thermo Scientific Chemicals
L05187-03
L05187-06
CAAAAL05187-03EA
114.87
CAD
CAAAAL05187-03
CAAAAL05187-06
11-Heneicosanol 97%
11-Heneicosanol
Purity: 97%
CAS No.: 3381-26-8
Molecular Formula: C21H44O
Formula Weight: 312.58
Odor: Odorless
Melting Point: 71-73°
Beilstein Reference: 1765841
MDL No.: MFCD00026547
Formula:
C₂₁H₄₄O Melting Pt: 71…73 °C Storage Temperature: Ambient |
MDL Number:
MFCD00026547 CAS Number: 3381-26-8 EINECS: 222-184-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...