Glyoxime ≥98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L05130-09
L05130-18
CAAAAL05130-09EA
107.91
CAD
CAAAAL05130-09
CAAAAL05130-18
Glyoxime ≥98%
Glyoxime
Purity: 98+%
CAS No.: 557-30-2
Molecular Formula: C2H4N2O2
Formula Weight: 88.07
Melting Point: ca 169° dec.
Beilstein Reference: 1699539
UN No.: 2926
MDL No.: MFCD00013942
Formula:
C₂H₄N₂O₂ Melting Pt: ca 169 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00013942 CAS Number: 557-30-2 EINECS: 209-168-1 UN: 2926 ADR: "4.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...