N-Phenylurethane 98%

Supplier: Thermo Scientific Chemicals
Warning

L05027-14 L05027-22
CAAAAL05027-14EA 90.69 CAD
CAAAAL05027-14 CAAAAL05027-22
N-Phenylurethane 98%
N-Phenylurethane

Purity: 98% CAS No.: 101-99-5 Molecular Formula: C6H5NHCO2CH2CH3 Formula Weight: 165.19 Refractive Index: 1.5375 Melting Point: 49-51° Boiling Point: 237° Beilstein Reference: 1942785 Merck Reference: 14,7320 MDL No.: MFCD00026806

Formula: C₉H₁₁NO₂
Boiling Pt: 237 °C
Melting Pt: 49…51 °C
Density: 1.106
Storage Temperature: Ambient
MDL Number: MFCD00026806
CAS Number: 101-99-5
EINECS: 202-995-9
Merck Index: 14,7320

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR