Cinnamaldoxime mixture E/Z 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L05026-06
L05026-14
CAAAAL05026-06EA
57.2
CAD
CAAAAL05026-06
CAAAAL05026-14
Cinnamaldoxime mixture E/Z 98%
Cinnamaldoxime
Purity: 98%
CAS No.: 13372-81-1
Molecular Formula: C9H9NO
Formula Weight: 147.18
Melting Point: 117-122°
Beilstein Reference: 3117555
MDL No.: MFCD00019969
Formula:
C₉H₉NO Melting Pt: 117…122 °C Storage Temperature: Ambient |
MDL Number:
MFCD00019969 CAS Number: 13372-81-1 EINECS: 236-449-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...