L-Adrenaline ≥98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
L-Epinephrine
L04911-06
L04911-14
CAAAAL04911-06EA
125.29
CAD
CAAAAL04911-06
CAAAAL04911-14
L-Adrenaline ≥98%
L-Adrenaline
Purity: 98+%
CAS No.: 51-43-4
Molecular Formula: C9H13NO3
Formula Weight: 183.20
Storage Temperature: 2°C to 8°C
Melting Point: 216-219°
Beilstein Reference: 2368277
Optical Rotation: -52° (c=1 in 0.6N HCl)
UN No.: 2811
MDL No.: MFCD00002204
Formula:
C₉H₁₃NO₃ MW: 183.21 g/mol Melting Pt: 216…219 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00002204 CAS Number: 51-43-4 EINECS: 200-098-7 UN: 2811 ADR: 6.1 III Merck Index: 14,3619 |
Specification Test Results
Optical rotation | -52° (c=1 in 0.6N HCl) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...