N-Methylpentylamine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
N-Pentylmethylamine
L04882-04
L04882-09
CAAAAL04882-04EA
143.62
CAD
CAAAAL04882-04
CAAAAL04882-09
N-Methylpentylamine 98%
N-Methylpentylamine
Purity: 98%
CAS No.: 25419-06-1
Molecular Formula: CH3(CH2)4NHCH3
Formula Weight: 101.19
Flash Point: 18°(64°F)
Refractive Index: 1.4105
Boiling Point: 117-119°
Beilstein Reference: 1731333
UN No.: 2733
MDL No.: MFCD00041354
Formula:
C₆H₁₅N MW: 101.19 g/mol Boiling Pt: 117…119 °C Density: 0.738 Flash Pt: 18 °C (64 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041354 CAS Number: 25419-06-1 EINECS: 246-966-9 UN: 2733 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...