2-(2-Thienyl)pyridine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-(Thiophen-2-yl)pyridine, 2-(2-Pyridyl)thiophene
L04775-03
L04775-06
CAAAAL04775-03EA
97.17
CAD
CAAAAL04775-03
CAAAAL04775-06
2-(2-Thienyl)pyridine 97%
2-(2-Thienyl)pyridine
Purity: 97%
CAS No.: 3319-99-1
Molecular Formula: C9H7NS
Formula Weight: 161.23
Melting Point: 60-65°
Beilstein Reference: 508454
MDL No.: MFCD00044441
Formula:
C₉H₇NS MW: 161.22 g/mol Melting Pt: 60…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00044441 CAS Number: 3319-99-1 EINECS: 222-022-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...