N,N-Diisopropylethylenediamine 97%

Supplier: Thermo Scientific Chemicals
Danger

L04765-09 L04765-18
CAAAAL04765-09EA 79.32 CAD
CAAAAL04765-09 CAAAAL04765-18
N,N-Diisopropylethylenediamine 97%
N,N-Diisopropylethylenediamine

Purity: 97% CAS No.: 121-05-1 Molecular Formula: C8H20N2 Formula Weight: 144.26 Flash Point: 54°(129°F) Refractive Index: 1.4420 Boiling Point: 170° Beilstein Reference: 1734800 UN No.: 2733 MDL No.: MFCD00015015

Formula: C₈H₂₀N₂
Boiling Pt: 170 °C
Density: 0.828
Flash Pt: 54 °C(129 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015015
CAS Number: 121-05-1
EINECS: 204-447-4
UN: 2734
ADR: "8",II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR