(±)-α-Lipoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-(1,2-Dithiolan-3-yl)pentanoic acid, (±)-1,2-Dithiolane-3-pentanoic acid, 5-(1,2)Dithiolan-3-yl-pentanoic acid, DL-α-Lipoic acid, DL-Thioctic acid (oxidized), DL-6,8-Thioctic acid, Lip(S2), 6,8-Dithiooctanoic acid
Alpha Lipoic acid. A hydrogen transferrin cofactor.
Purity: 98%
CAS No.: 1077-28-7
Molecular Formula: C8H14O2S2
Formula Weight: 206.33
Storage Temperature: 2°C to 8°C
Melting Point: 59-63°
Boiling Point: 160-165°
Beilstein Reference: 81853
Merck Reference: 14,9326
MDL No.: MFCD00005474
Formula:
C₈H₁₄O₂S₂ MW: 206.33 g/mol Boiling Pt: 160…165 °C Melting Pt: 59…63 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005474 CAS Number: 1077-28-7 EINECS: 214-071-2 Merck Index: 14,9326 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...