(R)-Ethyl-2-amino-3-mercaptopropanoate hydrochloride ≥98%

Supplier: Thermo Scientific Chemicals

Synonyms: L-Cysteine ethyl ester hydrochloride, H-Cys-Oet.HCl, H-Cys-OEt·HCl

L04692-14 L04692-22
CAAAAL04692-14EA 78.68 CAD
CAAAAL04692-14 CAAAAL04692-22
(R)-Ethyl-2-amino-3-mercaptopropanoate hydrochloride ≥98%
(R)-Ethyl-2-amino-3-mercaptopropanoate hydrochloride

Purity: 98+% CAS No.: 868-59-7 Molecular Formula: C5H11NO2S.HCl Formula Weight: 185.67 Melting Point: 123-125° Beilstein Reference: 3562600 Optical Rotation: -12° (c=1 in 1N HCl) MDL No.: MFCD00012631

Formula: C₅H₁₁NO₂S·HCl
MW: 185.666 g/mol
Melting Pt: 123…125 °C
Storage Temperature: Ambient
MDL Number: MFCD00012631
CAS Number: 868-59-7
EINECS: 212-779-6

Order Now

Specification Test Results

Optical rotation -12° (c=1 in 1N HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR